Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Certified Reference Materials, Oxide, Rocklabs

OxJ Oxide, 2.00 - 3.00 ppm Au, 50 g, Sachet
Catalog # CA10067-922
Supplier:   ROCKLABS (W/T) CA
 :  OXJ-AU-100X50G
Certified Reference Materials, Oxide, Rocklabs
Catalog # CA10067-922
Supplier:  ROCKLABS (W/T) CA
CAS Number:  
Certified Reference Materials, Oxide, Rocklabs

OxJ Oxide, 2.00 - 3.00 ppm Au, 50 g, Sachet

CAS Number:  
Price
$2,599.84
Log in for availability
Pack of 100 (50g)
Product Details & Documents

Rocklabs Gold Standards are reference materials that are certified for either Pt, Pd, Au or Ag

Recommended Gold (ppm) concentrations. Packaged in jars or satchets.

Specifications

Specifications

  • Pack type:
    Sachet
  • Description:
    OxJ Oxide, 2.00 - 3.00 ppm Au
  • Size:
    50 g

Specifications