Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Certified Reference Materials, Oxide, Rocklabs

OxQ Oxide, 20.00 - 50.00 ppm Au, 30 g, Sachet
Catalog # CA10067-892
Supplier:   ROCKLABS (W/T) CA
 :  OXQ-AU-100X30G
Certified Reference Materials, Oxide, Rocklabs
Catalog # CA10067-892
Supplier:  ROCKLABS (W/T) CA
CAS Number:  
Certified Reference Materials, Oxide, Rocklabs

OxQ Oxide, 20.00 - 50.00 ppm Au, 30 g, Sachet

CAS Number:  
Price
$2,224.41
Log in for availability
Pack of 100 (30g)
Product Details & Documents

Rocklabs Gold Standards are reference materials that are certified for either Pt, Pd, Au or Ag

Recommended Gold (ppm) concentrations. Packaged in jars or satchets.

Specifications

Specifications

  • Pack type:
    Sachet
  • Description:
    OxQ Oxide, 20.00 - 50.00 ppm Au
  • Size:
    30 g

Specifications