Order Entry
Canada
Orders LinkContactUsLinkComponent
Certified Reference Materials, Oxide, Rocklabs

Certified Reference Materials, Oxide, Rocklabs

OxN Oxide, 7.00 - 10.00 ppm Au, 30 g, Sachet
Catalog # CA10067-884
Supplier:   ROCKLABS (W/T) CA
Supplier #:  OXN-AU-100X30G
Certified Reference Materials, Oxide, Rocklabs
Catalog # CA10067-884
Supplier:  ROCKLABS (W/T) CA
CAS Number:  
Certified Reference Materials, Oxide, Rocklabs

OxN Oxide, 7.00 - 10.00 ppm Au, 30 g, Sachet

CAS Number:  
Price
$2,224.41
Log in for availability
Pack of 100 (30g)
Product Details & Documents

Rocklabs Gold Standards are reference materials that are certified for either Pt, Pd, Au or Ag

Recommended Gold (ppm) concentrations. Packaged in jars or satchets.

Specifications

Specifications

  • Pack type:
    Sachet
  • Description:
    OxN Oxide, 7.00 - 10.00 ppm Au
  • Size:
    30 g

Specifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.