Order Entry
Canada
Orders LinkContactUsLinkComponent
Certified Reference Materials, Oxide, Rocklabs

Certified Reference Materials, Oxide, Rocklabs

OxK Oxide, 3.00 - 5.00 ppm Au, 100 g, Sachet
Catalog # CA10067-820
Supplier:   ROCKLABS (W/T) CA
Supplier #:  OXK-AU-100X100G
Certified Reference Materials, Oxide, Rocklabs
Catalog # CA10067-820
Supplier:  ROCKLABS (W/T) CA
CAS Number:  
Certified Reference Materials, Oxide, Rocklabs

OxK Oxide, 3.00 - 5.00 ppm Au, 100 g, Sachet

CAS Number:  
Price
$4,950.94
Log in for availability
Pack of 100 (100g)
Product Details & Documents

Rocklabs Gold Standards are reference materials that are certified for either Pt, Pd, Au or Ag

Recommended Gold (ppm) concentrations. Packaged in jars or satchets.

Specifications

Specifications

  • Pack type:
    Sachet
  • Description:
    OxK Oxide, 3.00 - 5.00 ppm Au
  • Size:
    100 g

Specifications