Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Certified Reference Materials, Oxide, Rocklabs

OxE Oxide, 0.50 - 0.70 ppm Au, 100 g, Sachet
Catalog # CA10067-798
Supplier:   ROCKLABS (W/T) CA
 :  OXE-AU-100X100G
Certified Reference Materials, Oxide, Rocklabs
Catalog # CA10067-798
Supplier:  ROCKLABS (W/T) CA
CAS Number:  
Certified Reference Materials, Oxide, Rocklabs

OxE Oxide, 0.50 - 0.70 ppm Au, 100 g, Sachet

CAS Number:  
Price
4 950,94 $
Log in for availability
Pack of 100 (100g)
Product Details & Documents

Rocklabs Gold Standards are reference materials that are certified for either Pt, Pd, Au or Ag

Recommended Gold (ppm) concentrations. Packaged in jars or satchets.

Specifications

Specifications

  • Pack type:
    Sachet
  • Description:
    OxE Oxide, 0.50 - 0.70 ppm Au
  • Size:
    100 g

Specifications