Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Certified Reference Materials, Oxide, Rocklabs

OxD Oxide, 0.30 - 0.50 ppm Au, 100 g, Sachet
  CA10067-796
 :  OXD-AU-100X100G
Certified Reference Materials, Oxide, Rocklabs
  CA10067-796
 :  
Certified Reference Materials, Oxide, Rocklabs

OxD Oxide, 0.30 - 0.50 ppm Au, 100 g, Sachet

 :  
 
$4,950.94
   
Pack of 100 (100g)
 

Rocklabs Gold Standards are reference materials that are certified for either Pt, Pd, Au or Ag

Recommended Gold (ppm) concentrations. Packaged in jars or satchets.

 

 

  • Pack type:
    Sachet
  • Description:
    OxD Oxide, 0.30 - 0.50 ppm Au
  • Size:
    100 g