Orders LinkContactUsLinkComponent
TempDex 710, Nitrile Gloves, Mapa Professional
TempDex 710, Nitrile Gloves, Mapa Professional
9, Yellow, 26 cm (10³⁄₁₆")
Catalog # 10067-642
Supplier:   MAPA
Supplier #:  34710039
TempDex 710, Nitrile Gloves, Mapa Professional
Catalog # 10067-642
Supplier:  MAPA
TempDex 710, Nitrile Gloves, Mapa Professional
TempDex 710, Nitrile Gloves, Mapa Professional
9, Yellow, 26 cm (10³⁄₁₆")
Price
$13.82
New Customer Promotional Price
Log in for availability
Each Case of 50
Product Details & Documents
These high dexterity and thermal protection gloves are suitable for handling of composite materials after heat processing in ceramics, plastics and composite industries.
  • Nitrile coated palm and dotted embossing for better insulation and grip
  • Thin, anatomically-shaped glove ensures dexterity
  • Long lasting and abrasion resistant
Ideal for use in ceramics, plastics, composite and construction industries.
Specifications
  • Glove color:
    Yellow
  • Glove size:
    9
  • Length:
    26 cm (10³⁄₁₆")