Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Certified Reference Materials, Oxide, Rocklabs

OxE Oxide, 0.50 - 0.70 ppm Au, 2.5 kg, Jar
  CA10067-782
 :  OXE-AU-2.5KG
Certified Reference Materials, Oxide, Rocklabs
  CA10067-782
 :  
Certified Reference Materials, Oxide, Rocklabs

OxE Oxide, 0.50 - 0.70 ppm Au, 2.5 kg, Jar

 :  
 
$380.88
   
Each (2,500g)
 

Rocklabs Gold Standards are reference materials that are certified for either Pt, Pd, Au or Ag

Recommended Gold (ppm) concentrations. Packaged in jars or satchets.

 

 

  • Pack type:
    Jar
  • Description:
    OxE Oxide, 0.50 - 0.70 ppm Au
  • Size:
    2.5 kg