Order Entry
Canada
Orders LinkContactUsLinkComponent
Caffeine

Caffeine

Caffeine, Laboratory, 10 g
Catalog # 470300-520
Supplier:   Ward's Science
Supplier #:  CC0025-10G
Caffeine
Catalog # 470300-520
Supplier:  Ward's Science
CAS Number:  
Caffeine
Caffeine

Caffeine, Laboratory, 10 g

CAS Number:  
Price
23,15 $
Log in for availability
Each (10g)
Minimum order quantity is 1
Product Details & Documents

CAS Number: 58-08-2
Formula Weight: 194.20
Formula: C8H10N4O2
Hazard Info: Toxic
Density (g/mL): 1.23
Boiling Point (°C): 178
Freezing Point (°C): 238
Solubility: Chloroform, Water and Alcohol
Synonyms: 1,3,7-Trimethylxanthine
Shelf Life (months): 12
Storage: Blue

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Specifications

Specifications

  • Description:
    Caffeine
  • Size:
    10 g
  • Chemical/physical specifications:
    Powder
  • Grade:
    Laboratory

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.