Order Entry
Canada
Orders LinkContactUsLinkComponent
Barium Nitrate

Barium Nitrate

Barium Nitrate, Reagent, 100 g
Catalog # 470300-346
Supplier:   Ward's Science
Supplier #:  BB0055-100GR
Barium Nitrate
Catalog # 470300-346
Supplier:  Ward's Science
CAS Number:  
Barium Nitrate
Barium Nitrate

Barium Nitrate, Reagent, 100 g

CAS Number:  
Price
$167.55
Log in for availability
Each (100g)
Minimum order quantity is 1
Product Details & Documents

CAS Number: 10022-31-8
Formula Weight: 261.35
Formula: Ba(NO3)2
Hazard Info: Oxidizer, Toxic
Density (g/mL): 3.24
Boiling Point (°C): Decomposes
Freezing Point (°C): 592
Solubility: Water
Synonyms: Barium Dinitrate, Nitric Acid Barium Salt
Shelf Life (months): 36
Storage: Yellow

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Specifications

Specifications

  • Description:
    Barium Nitrate
  • Size:
    100 g
  • Chemical/physical specifications:
    Crystal
  • Grade:
    Reagent

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.