Order Entry
Canada
Orders LinkContactUsLinkComponent
Potassium Nitrate

Potassium Nitrate

Potassium Nitrate, Reagent, 500 g
Catalog # 470302-204
Supplier:   Ward's Science
Supplier #:  PP0655-500GR
Potassium Nitrate
Catalog # 470302-204
Supplier:  Ward's Science
CAS Number:  
Potassium Nitrate
Potassium Nitrate

Potassium Nitrate, Reagent, 500 g

CAS Number:  
Price
$34.60
Log in for availability
Each (500g)
Minimum order quantity is 1
Product Details & Documents

CAS Number: 7757-79-1
Formula: KNO3
Density: 2.1 g/mL
Boiling and Freezing Point: 400°C, 333°C
Solubility: Water, Glycerin, and Alcohol
Synonyms: Potash Nitrate, Saltpeter
Shelf Life: 36 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Caution: This product is flammable oxidizer and irritant.

Specifications

Specifications

  • Description:
    Potassium Nitrate
  • Size:
    500 g
  • Chemical/physical specifications:
    Crystal
  • Grade:
    Reagent

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.