Potassium Nitrate
Potassium Nitrate, Laboratory, 500 g
Supplier: Ward's Science
: PP0650-500G
Jump to:
Product Details & Documents
CAS Number: 7757-79-1
Formula: KNO3
Density: 2.1 g/mL
Boiling and Freezing Point: 400°C, 333°C
Solubility: Water, Glycerin, and Alcohol
Synonyms: Potash Nitrate, Saltpeter
Shelf Life: 36 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
: This product is flammable oxidizer and irritant.
Specifications
Specifications
- Description:Potassium Nitrate
- Size:500 g
- Chemical/physical specifications:Prill
- Grade:Laboratory



