Order Entry
Canada
Orders LinkContactUsLinkComponent
Phosphate Buffer

Phosphate Buffer

Phosphate Buffer, Solution pH 6.5, Laboratory, 1 L
Catalog # 470302-014
Supplier:   Ward's Science
Supplier #:  PP0235-1L
Phosphate Buffer
Catalog # 470302-014
Supplier:  Ward's Science
CAS Number:  
Phosphate Buffer
Phosphate Buffer

Phosphate Buffer, Solution pH 6.5, Laboratory, 1 L

CAS Number:  
Price
$27.75
Log in for availability
Each (1l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Caution: Please Note: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Specifications

Specifications

  • Description:
    Phosphate Buffer, Solution pH 6.5
  • Size:
    1 L
  • Chemical/physical specifications:
    0.1 M
  • Grade:
    Laboratory

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.