Order Entry
Canada
Orders LinkContactUsLinkComponent
Nucleofector® P2 Primary Cell 4D-X Kit L

Nucleofector® P2 Primary Cell 4D-X Kit L

P2 primary cell 4D-Nucleofector® X kit L, 12
Catalog # CA10064-236
Supplier #:  V4XP-2012
Nucleofector® P2 Primary Cell 4D-X Kit L
Catalog # CA10064-236
CAS Number:  
Nucleofector® P2 Primary Cell 4D-X Kit L
Nucleofector® P2 Primary Cell 4D-X Kit L

P2 primary cell 4D-Nucleofector® X kit L, 12

CAS Number:  
Price
$590.52
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The P2 Primary Cell 4D-Nucleofector® X kit L is one of our five 4D-Nucleofector® kits suited for transfecting primary cells in the 100 µl Nucleocuvette® vessel format when working with the 4D-Nucleofector® X unit.

  • High transfection efficiencies for primary cells
  • Cells maintain functionality post transfection
  • Conditions are transferable between 4D-Nucleofector® System, 96-well Shuttle® System and 384-well Nucleofector® System

This kit has been determined to be optimal for transfecting following cell types:
Macrophage (mouse)
NHDF (human), neonatal

Applications:
Transfection of cell numbers in the range of 2×10⁵ to 2×10⁷ cells.
Low throughput applications.

The kit is available in different variants for either 12 or 24 reactions.

 : CA10064-236 includes 2×0.675 ml P1 primary cell Nucleofector® solution, 2×0.15 ml supplement 1, 50 µg pmaxGFP® Vector (1 μg/μl in 10 mM Tris pH 8.0), 12×100 µl Nucleocuvette® vessel, 12×single use pipettes.

CA10064-238 includes 2.25 ml P1 Primary Cell Nucleofector® solution, 0.5 ml supplement 1, 50 µg pmaxGFP® Vector (1 μg/μl in 10 mM Tris pH 8.0), 24×100 µl Nucleocuvette® vessel, 24×single use pipettes.

Specifications

Specifications

  • Description:
    P2 primary cell 4D-Nucleofector® X kit L
  • No. of reactions:
    12

Specifications