Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Ammonium Hydroxide

Ammonium hydroxide, solution, Reagent ACS, 500 ml
  470300-212
 :  AA0264-500MLR
Ammonium Hydroxide
  470300-212
 :  
Ammonium Hydroxide
Ammonium Hydroxide

Ammonium hydroxide, solution, Reagent ACS, 500 ml

 :  
 
$36.10
   
Each (500ml)
 
 

CAS Number: 1336-21-6
Formula: NH4OH
Density: 0.900 g/mL
Boiling and Freezing Point: 36°C, -77°C
Synonyms: Ammonium Hydroxide Water Solution
Shelf Life: 36 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

 : This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

 : Corrosive, irritant and toxic.

 

 

  • Description:
    Ammonium hydroxide, solution
  • Size:
    500 ml
  • Chemical/physical specifications:
    29% (14.8 M)
  • Grade:
    Reagent ACS