Order Entry
Canada
Orders LinkContactUsLinkComponent
Calcium Hydroxide

Calcium Hydroxide

Calcium Hydroxide, Laboratory, 500 g
Catalog # 470300-588
Supplier:   Ward's Science
Supplier #:  CC0095-500G
Calcium Hydroxide
Catalog # 470300-588
Supplier:  Ward's Science
CAS Number:  
Calcium Hydroxide
Calcium Hydroxide

Calcium Hydroxide, Laboratory, 500 g

CAS Number:  
Price
$22.05
Log in for availability
Each (500g)
Minimum order quantity is 1
Product Details & Documents

CAS Number: 1305-62-0
Formula Weight: 74.10
Formula: Ca(OH)2
Hazard Info: Irritant, Corrosive
Density (g/mL): 2.24
Boiling Point (°C): as CaO 579
Freezing Point (°C): as CaO 2580
Solubility: Water, Glycerol, and Acids
Synonyms: Hydrated Lime, Slacked Lime, Caustic Lime, Calcium Hydrate
Shelf Life (months): 12
Storage: Green

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Specifications

Specifications

  • Description:
    Calcium Hydroxide
  • Size:
    500 g
  • Chemical/physical specifications:
    Powder
  • Grade:
    Laboratory

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.