Sodium Hydroxide
Sodium Hydroxide, Reagent, 2.5 kg
: SS0550-2.5KGR
CAS Number: 1310-73-2
Formula: NaOH
Density: 2.13 g/mL
Boiling and Freezing Point: 1390°C, 318°C
Solubility: Water and Alcohol
Synonyms: Caustic Soda
Shelf Life: 36 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
: This product is corrosive and toxic.
- Description:Sodium Hydroxide
- Size:2.5 kg
- Chemical/physical specifications:Pellet
- Grade:Reagent



