Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
Tan
Catalog # 490010-384
Supplier #: T312-8
Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
Catalog # 490010-384
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
CAPINSERT F/CRYOVIAL TAN 500CS Catalog #CA73960-401
Product CA73960-401 is a replacement for 490010-384 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Color coded inserts fit precisely into the cap of the Cryovial® tubes for color identification.
Made of polypropylene
Ordering information: T312-7 contains 100 pieces each of white, blue, red, green and yellow inserts packaged in individual bags.
Documents
Specifications
- Color:Tan