Orders LinkContactUsLinkComponent
Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
Yellow
Catalog # 490010-380
Supplier #:  T312-5
Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
Catalog # 490010-380
Supplier:  
Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
Yellow
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
INSERT CRYOVIAL - YLW 500/PK Catalog #CA73960-406
Product CA73960-406 is a replacement for 490010-380 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Color coded inserts fit precisely into the cap of the Cryovial® tubes for color identification.
Made of polypropylene
Ordering information: T312-7 contains 100 pieces each of white, blue, red, green and yellow inserts packaged in individual bags.
Specifications
  • Color:
    Yellow