Order Entry
Canada
Orders LinkContactUsLinkComponent
PC Link Labels, Brady

PC Link Labels, Brady

Labels for portable thermal printing, Polyester, White/translucent, TLS 2200®, TLS PC Link®, BMP®71, BMP®61, 0.75×3.75"
Catalog # 490006-246
PC Link Labels, Brady
Catalog # 490006-246
Supplier:  
PC Link Labels, Brady
PC Link Labels, Brady

Labels for portable thermal printing, Polyester, White/translucent, TLS 2200®, TLS PC Link®, BMP®71, BMP®61, 0.75×3.75"

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
TLS PRINTER LABELS B-461 MAT'L Catalog #CA80090-148

Product CA80090-148 is a replacement for 490006-246 and has the same packaging and specifications, but may be a different quantity.

Product Details & Documents

B-461: Self-laminating polyester is a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
B-490: Durable polyester markers are designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.

B-499: Nylon cloth wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
B-423: Permanent polyester is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.

B-427: Self-laminating vinyl is an excellent material for wire and cable identification. This material has good clarity, conformability and is self-extinguishing.
B-351: Tamper-resistant vinyl is designed to fracture easily to show signs of product tampering and to prevent on-piece label removal.

B-435: Glossy metallized permanent polyester is designed for high adhesion to textured metals, powder coated surfaces and low surface energy plastics.

B-488: High performance polyester withstands numerous solvents and variable temperatures when applied to various surfaces including stainless steel and polypropylene.

Specifications
  • For Use With:
    TLS 2200®, TLS PC Link®, BMP®71, BMP®61
  • Color:
    White/translucent
  • Description:
    Labels for portable thermal printing
  • Dimensions:
    0.75×3.75"
  • Material:
    Polyester
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.