Order Entry
Canada
Orders LinkContactUsLinkComponent
Cryogenic Printer Labels, Brady

Cryogenic Printer Labels, Brady

6200 series thermal transfer printer ribbon, IP™ printer enabled, Black, IP300®, IP600®, 2.36", 300 m (984 ft.)
Catalog # 490006-112
Cryogenic Printer Labels, Brady
Catalog # 490006-112
Supplier:  
Cryogenic Printer Labels, Brady
Cryogenic Printer Labels, Brady

6200 series thermal transfer printer ribbon, IP™ printer enabled, Black, IP300®, IP600®, 2.36", 300 m (984 ft.)

Discontinued
Product Details & Documents

Cryogenic labels for tubes, vials, and canes.

Specifications
  • For Use With:
    IP300®, IP600®
  • Color:
    Black
  • Description:
    6200 series thermal transfer printer ribbon, IP™ printer enabled
  • Roll length:
    300 m (984 ft.)
  • Width:
    2.36"