Orders LinkContactUsLinkComponent
Cell Strainers
Cell Strainers
70 µm, White
Catalog # 10054-456
Supplier:   Corning
Supplier #:  431751
Cell Strainers
Catalog # 10054-456
Supplier:  Corning
Cell Strainers
Cell Strainers
70 µm, White
Price
$209.40
New Customer Promotional Price
Log in for availability
Case of 50
Product Details & Documents
Corning® cell strainers are sterile, easy to use devices for rapidly isolating primary cells to consistently obtain a uniform single-cell suspension from tissues. Fits 50 ml Falcon® conical tubes.
  • Strong nylon mesh for optimal performance in a variety of applications
  • Extended lip on strainer enables aseptic handling with forceps
  • Moulded color-coded polypropylene frame with tab enables easy handling and identification
  • Sterilised by gamma irradiation, non cytotoxic
  • Conveniently accessible in individual packaging
Use these versatile cell strainers to prepare samples for flow cytometry, single cell suspensions of blood cells, specimens for primary cell cultures and immunogens, as well as primary cells from tissues.
Specifications
  • Pore size:
    70 µm
  • Color:
    White