Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SATWIPES* PET 9X11 70/30 IPA/DI CS12
Catalog # 10053-744
Supplier:
Contec
SDS
Expand image
SATWIPES* PET 9X11 70/30 IPA/DI CS12
Catalog # 10053-744
Supplier:
Contec
Supplier Number:
SWC33120
SDS
Related Information
Documentation
SDS
GHS_SWC33120_07232014
Additional Product Details
VWR GHS Compliance Letter (US)
Documentation
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International