Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PERFECTPRO NI-NTA SUPERFLOW 10ML
Catalog # 10052-184
Supplier:
Quantabio
SDS
Expand image
PERFECTPRO NI-NTA SUPERFLOW 10ML
Catalog # 10052-184
Supplier:
Quantabio
Supplier Number:
2400060
SDS
Related Information
Documentation
SDS
2400060_02272013
Additional Product Details
VWR GHS Compliance Letter (US)
Documentation
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International