$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
PHASE LOCK GEL HEAVY 2ML EA=200TUBES
10052-170
:
Quantabio
PHASE LOCK GEL HEAVY 2ML EA=200TUBES
10052-170
:
Quantabio
:
2302830
Phase Lock Gel Datasheet
Protocol Isolation of Genomic DNA from Paraffin-embedded sections
Protocol Isolation of Genomic DNA from Staphylococcus aureus
Protocol Maxi-prep of Genomic DNA from Leaf Tissue
Protocol RNA from Fatty Tissue
Phase Lock Gel Manual
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International