Orders LinkContactUsLinkComponent
Standard For Osmometer, Advanced Instruments
Standard For Osmometer, Advanced Instruments
1000 mOsm Calibration Standard, 10×2 ml
  CA89369-186
Supplier #:  3MA100
Standard For Osmometer, Advanced Instruments
  CA89369-186
 :  
Standard For Osmometer, Advanced Instruments
1000 mOsm Calibration Standard, 10×2 ml
 :  
$402.56
Pack of 10
Calibration standard is used to check the perforance of osmometer and test methods.
Documents
  • Description:
    1000 mOsm Calibration Standard
  • Size:
    10×2 ml
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)