Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Certified Reference Materials, Oxide, Rocklabs

OxJ Oxide, 2.00 - 3.00 ppm Au, 2.5 kg, Jar
Catalog # CA10032-320
Supplier:   ROCKLABS (W/T) CA
 :  OXJ-AU-2.5KG
Certified Reference Materials, Oxide, Rocklabs
Catalog # CA10032-320
Supplier:  ROCKLABS (W/T) CA
CAS Number:  
Certified Reference Materials, Oxide, Rocklabs

OxJ Oxide, 2.00 - 3.00 ppm Au, 2.5 kg, Jar

CAS Number:  
Price
$380.88
Log in for availability
Each (2,500g)
Product Details & Documents

Rocklabs Gold Standards are reference materials that are certified for either Pt, Pd, Au or Ag

Recommended Gold (ppm) concentrations. Packaged in jars or satchets.

Specifications

Specifications

  • Pack type:
    Jar
  • Description:
    OxJ Oxide, 2.00 - 3.00 ppm Au
  • Size:
    2.5 kg

Specifications