$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
PERFECTA MULTIPLEX TOUGHMIX LOW ROX1000R
CA11033-420
:
Quantabio
PERFECTA MULTIPLEX TOUGHMIX LOW ROX1000R
CA11033-420
:
Quantabio
:
95149-01K
ghs_95149
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International