Order Entry
Canada
Orders LinkContactUsLinkComponent
Certified Reference Materials, Oxide, Rocklabs

Certified Reference Materials, Oxide, Rocklabs

OxP Oxide, 10.00 - 20.00 ppm Au, 2.5 kg, Jar
Catalog # CA10029-286
Supplier:   ROCKLABS (W/T) CA
Supplier #:  OXP-AU-2.5KG
Certified Reference Materials, Oxide, Rocklabs
Catalog # CA10029-286
Supplier:  ROCKLABS (W/T) CA
CAS Number:  
Certified Reference Materials, Oxide, Rocklabs

OxP Oxide, 10.00 - 20.00 ppm Au, 2.5 kg, Jar

CAS Number:  
Price
$380.88
Log in for availability
Each (2,500g)
Product Details & Documents

Rocklabs Gold Standards are reference materials that are certified for either Pt, Pd, Au or Ag

Recommended Gold (ppm) concentrations. Packaged in jars or satchets.

Specifications

Specifications

  • Pack type:
    Jar
  • Description:
    OxP Oxide, 10.00 - 20.00 ppm Au
  • Size:
    2.5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.