Dropper Bottles, Qorpak
30 ml, Glass, Clear, 24-400
Catalog # 16164-333
Supplier: Qorpak
Supplier #: 500523
Jump to:
Product Details & Documents
Supplied with Attached Glass Dropper Assembly.
- Square, clear bottles
- Ideal for dispensing small amounts of liquid
Dropper Bottles feature black polypropylene closures, rubber bulbs, and straight point glass dropper tubes.
Specifications
- Cap Size:24-400
- Capacity:30 ml
- Color:Clear
- Material:Glass