Split Specimen Collection Kits, Therapak®
Specimen Collection Split Bag Kit
Catalog # 10029-336
Supplier: Therapak, LLC
Supplier #: 40002
Jump to:
Product Details & Documents
For drugs of abuse testing applications requiring two samples to be collected from a single donor, the split specimen collection and transport kits are available in either a split bag kit or the same kit within a transport box.
Delivery information: Kits include 6.5 oz. beaker style collection cup with pour-spout and temperature strip, 2 - 60 mL sterile containers with sterility seal and a transport bag.
Specifications
- Description:Specimen Collection Split Bag Kit