Order Entry
Canada
Orders LinkContactUsLinkComponent
Pierce™ RIPA Buffer, Thermo Scientific

Pierce™ RIPA Buffer, Thermo Scientific

RIPA Lysis and Extraction Buffer, 100 mL
Catalog # CAPI89900
Supplier:   Thermo Scientific
Supplier #:  89900
Pierce™ RIPA Buffer, Thermo Scientific
Catalog # CAPI89900
Supplier:  Thermo Scientific
CAS Number:  
Pierce™ RIPA Buffer, Thermo Scientific
Pierce™ RIPA Buffer, Thermo Scientific

RIPA Lysis and Extraction Buffer, 100 mL

CAS Number:  
Price
$339.31
Log in for availability
Each (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Thermo Scientific RIPA Buffer is a high-quality, ready-to-use and fully disclosed formulation of a popular cell lysis reagent for cultured mammalian cells.

  • Compatible with many applications, including reporter assays, protein assays, immunoassays and protein purification
  • Enables extraction of cytoplasmic, membrane and nuclear proteins making it very versatile
  • Contains no proprietary components, providing users with complete control and knowledge of possible compatibility issues

This RIPA buffer effectively lyses and extracts protein from cultured mammalian cells, including plated cells and pelleted suspension cells.

RIPA buffer derives its name from the original application for which it was developed: the radio-immunoprecipitation assay. While this isotopic assay method is rarely performed in laboratories today, the acronym for this lysis buffer formulation has endured in common use. RIPA cell lysis reagent is highly effective for protein extraction from a variety of cell types because it contains three non-ionic and ionic detergents. One disadvantage of this detergent formulation is its relative incompatibility with certain downstream applications compared to other lysis reagents.

Specifications

Specifications

  • Description:
    RIPA Lysis and Extraction Buffer, 100 mL

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.