Order Entry
Canada
Orders LinkContactUsLinkComponent
Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific

Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific

TBS, 1 L
Catalog # CAPI37542
Supplier:   Thermo Scientific
Supplier #:  37542
Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific
Catalog # CAPI37542
Supplier:  Thermo Scientific
CAS Number:  
Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific
Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific

TBS, 1 L

CAS Number:  
Price
$634.33
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

These StartingBlock™ blocking buffers are PBS and TBS formulations of a proprietary protein that provides exceptional performance and broad compatibility with membrane (Western blot) and 96-well plate (ELISA) assays.

  • Compatible with many detection systems (Western blot, ELISA and IHC)
  • Short blocking times
  • High signal:noise ratio
  • Strip and reprobe without reblocking
  • Available ready to use 1X formulations in PBS and TBS

StartingBlock Blocking Buffers performs well with an extremely wide range of antibodies, antibody combinations, other protein probing and assay systems.

These buffers are biotin- and serum-free. They are compatible with many detection systems and feature short blocking times. These buffers also allow stripping and reprobing with reblocking to save time and money.

Caution: For Research Use Only. Not for use in diagnostic procedures. Upon receipt store at 4˚C.

Specifications

Specifications

  • Description:
    TBS
  • Size:
    1 L
  • Application:
    ELISA, Western Blot, Dot Blot, Immunohistochemistry

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.