Pierce™ StartingBlock™ (PBS) Blocking Buffer, Thermo Scientific
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
These StartingBlock™ blocking buffers are PBS and TBS formulations of a proprietary protein that provides exceptional performance and broad compatibility with membrane (Western blot) and 96-well plate (ELISA) assays.
- Compatible with many detection systems (Western blot, ELISA and IHC)
- Short blocking times
- High signal:noise ratio
- Strip and reprobe without reblocking
- Available ready to use 1X formulations in PBS and TBS
StartingBlock Blocking Buffers performs well with an extremely wide range of antibodies, antibody combinations, other protein probing and assay systems.
These buffers are biotin- and serum-free. They are compatible with many detection systems and feature short blocking times. These buffers also allow stripping and reprobing with reblocking to save time and money.
Caution: For Research Use Only. Not for use in diagnostic procedures. Upon receipt store at 4˚C.
Documents
Certificates
Search by Lot NumberThermo Scientific Reagents for Manual Western Blot Detection Brochure
Product Literature
Specifications
Specifications
- Description:PBS
- Size:1 L
- Application:ELISA, Western Blot, Dot Blot, Immunohistochemistry
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.