Order Entry
Canada
Orders LinkContactUsLinkComponent
Pierce™ Blocker™ BSA Blocking Buffer, Thermo Scientific

Pierce™ Blocker™ BSA Blocking Buffer, Thermo Scientific

Blocker™ BSA (10X) in TBS, Blocking Buffer, 125 mL
Catalog # CAPI37520
Supplier:   Thermo Scientific
Supplier #:  37520
Pierce™ Blocker™ BSA Blocking Buffer, Thermo Scientific
Catalog # CAPI37520
Supplier:  Thermo Scientific
CAS Number:  
Pierce™ Blocker™ BSA Blocking Buffer, Thermo Scientific
Pierce™ Blocker™ BSA Blocking Buffer, Thermo Scientific

Blocker™ BSA (10X) in TBS, Blocking Buffer, 125 mL

CAS Number:  
Price
$486.92
Log in for availability
Each (125ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Blocker BSA in PBS or TBS are ready-to-use, 1X PBS or TBS solutions of 10X (10%) bovine serum albumin protein for blocking steps in Western blot, ELISA, IHC and nucleic acid detection methods.

  • Easy to use
  • Convenient and saves storage space
  • Purified protein provides fewer chances of cross reaction with assay components

This 10% (w/v) solution of high quality BSA that is useful for saturating excess protein binding sites on membranes and microplates in immunoassays. Blocker BSA Buffers are most frequently diluted 10-fold (to 1% BSA) in 1X Tris-buffered saline for initial testing, but other buffer concentrations can be beneficial for specific systems. Blocker BSA is usually more effective than non fat milk blocking buffers for biotin-avidin systems because it contains a single purified protein that is devoid endogenous biotin.

Documents

Specifications

Specifications

  • Description:
    Blocker™ BSA (10X) in TBS, Blocking Buffer
  • Size:
    125 mL

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.