Order Entry
Canada
Orders LinkContactUsLinkComponent
 

SuperBlock™ (PBS) Blocking Buffer, Thermo Scientific

PBS Blotting, 1 L
  CAPI37517
 :  37517
SuperBlock™ (PBS) Blocking Buffer, Thermo Scientific
  CAPI37517
 :  
SuperBlock™ (PBS) Blocking Buffer, Thermo Scientific
SuperBlock™ (PBS) Blocking Buffer, Thermo Scientific

PBS Blotting, 1 L

 :  
 
$554.03
   
Each (1l)
 
   
 

SuperBlock™ Blocking Buffer is an optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis.

  • Blocking time—block membranes in 5 to 10 minutes and ELISA plates in 2 minutes
  • Applications—ELISA, western blotting, IHC; biotin-free for use with streptavidin system
  • Stability—store buffer at 4°C for one year; store blocked plates dry for up to 12 months

Blocking micro-plates, membranes or tissues with a SuperBlock Buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin, or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilising them for drying and storage for later use.

 

 

  • Description:
    PBS Blotting
  • Size:
    1 L
  • Application:
    Western Blot, Dot Blot, Immunohistochemistry

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.