Order Entry
Canada
Orders LinkContactUsLinkComponent
SuperBlock™ (PBS) Blocking Buffer, Thermo Scientific

SuperBlock™ (PBS) Blocking Buffer, Thermo Scientific

PBS, 1 L
Catalog # CAPI37515
Supplier:   Thermo Scientific
Supplier #:  37515
SuperBlock™ (PBS) Blocking Buffer, Thermo Scientific
Catalog # CAPI37515
Supplier:  Thermo Scientific
CAS Number:  
SuperBlock™ (PBS) Blocking Buffer, Thermo Scientific
SuperBlock™ (PBS) Blocking Buffer, Thermo Scientific

PBS, 1 L

CAS Number:  
Price
$617.94
Log in for availability
Each (1l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

SuperBlock™ Blocking Buffer is an optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis.

  • Blocking time—block membranes in 5 to 10 minutes and ELISA plates in 2 minutes
  • Applications—ELISA, western blotting, IHC; biotin-free for use with streptavidin system
  • Stability—store buffer at 4°C for one year; store blocked plates dry for up to 12 months

Blocking micro-plates, membranes or tissues with a SuperBlock Buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin, or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilising them for drying and storage for later use.

Specifications

Specifications

  • Description:
    PBS
  • Size:
    1 L
  • Application:
    ELISA, Western Blot, Dot Blot, Immunohistochemistry, Hybridization

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.