Model ASC-1 Ascorbic Acid Test Kit, Hach
Product CA89500-718 is a replacement for 89500-718 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Model ASC-1 Ascorbic Acid 100-Test Kit was developed in cooperation with beverage producers and uses the drop-count titration method to measure ascorbic acid (vitamin C)
Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
Utilizes drop count titration with a range of 10-200 mg/L. This kit utilizes drop-count titration simply and economically. It contains all required reagents and apparatus in a rugged carrying case for analysis in the field.
Ordering information: Sample measuring tube, flask, reagents, instructions and carrying case included.
Documents
ghs_2308100
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
Specifications
- Chemical method:Iodometric
- Range:10 - 200 mg/L ascorbic acid
- No. of tests:100
- Cat. No.:89500-718
- Platform:DCT
- Method:Drop Count Titration/ Iodometric