Orders LinkContactUsLinkComponent
SLIMSETTE™ Biopsy Processing and Embedding Cassettes, Simport Scientific
SLIMSETTE™ Biopsy Processing and Embedding Cassettes, Simport Scientific
White
Catalog # CA11031-884
Supplier:   Simport Scientific
Supplier #:  M510-2
SLIMSETTE™ Biopsy Processing and Embedding Cassettes, Simport Scientific
Catalog # CA11031-884
Supplier:  Simport Scientific
SLIMSETTE™ Biopsy Processing and Embedding Cassettes, Simport Scientific
SLIMSETTE™ Biopsy Processing and Embedding Cassettes, Simport Scientific
White
Price
$446.10
Log in for availability
Case of 1,500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These compact, easy-to-use cassettes incorporate a unique recessed cover, which allows more cassettes to be stacked
They are molded from acetal polymer to keep specimens safely submerged in liquid and are resistant to chemical actions of histological solvents. The 1mm mesh holes are specially designed to hold biopsy specimens, and the one-piece integral lid can be opened and closed as often as necessary. The anterior writing area is slanted at a 45° angle for use with certain types of cassette labeling instruments
Specifications
  • Color:
    White
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)