Orders LinkContactUsLinkComponent
Eisco® Joint Models
Knee Joint
  470212-428
 :   Wards
 :  AM0134AS
Eisco® Joint Models
  470212-428
 :  Wards
Eisco® Joint Models
Eisco® Joint Models
Knee Joint
$164.00
Each
Study How Joints Are Held Together.
  • Shows ligaments of 4 common joints
  • Ligaments are slightly flexible, and joints are reinforced to minimize damage
  • Mounted on base
These joint models from Eisco are a great addition to your anatomy lessons. Joint structure is easily visible with both bone and ligaments visible. The ligament material has some resilience, and the joints are reinforced to reduce the chance of damage to the model while using. This limits the range of motion for each joint, but still allows your students to see how movement is transferred through the ligaments.
  • Description:
    Knee Joint