Orders LinkContactUsLinkComponent
Dropper Bottles
Dropper Bottles
Dropper Bottles, 4 ml
Catalog # 470211-800
Supplier #:  2752-9125
Dropper Bottles
Catalog # 470211-800
Dropper Bottles
Dropper Bottles
Dropper Bottles, 4 ml
Price
$119.00
Log in for availability
Case of 25
Product Details & Documents
These bottles are made of low-density polyethylene.
  • Bottles Dispense about 40 mcL per Drop
  • Good Chemical Resistance
  • Semi-Transparent Bottles
  • Polyethylene Construction
The semi-transparent bottles feature color-coded caps.
They have good chemical resistance and dispense about 40 mcL per drop.
Specifications
  • Capacity:
    4 ml
  • Description:
    Dropper Bottles