Order Entry
Canada
Orders LinkContactUsLinkComponent
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning

PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning

125 ml, 2
Catalog # 470211-470
Supplier:   Corning
Supplier #:  4060-125
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
Catalog # 470211-470
Supplier:  Corning
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning

125 ml, 2

Price
$339.00
Log in for availability
Pack of 6 Case of 24
Product Details & Documents

Manufactured from borosilicate glass.

Tooled mouth ensures uniform stopper fit. Florence, long neck. Normal working temperature is 230 °C (446 °F).

Documents

Specifications
  • Capacity:
    125 ml
  • Stopper no.:
    2