PYREX® Burets
Geissler Buret Locking Stopcock
Supplier: Corning
Supplier #: 2116-LSO
Jump to:
Product Details & Documents
Educational grade Pyrex burets.
- Heavy duty
- Transparent
Supplied with self-lubricating, leakproof PMP stopcock. This buret is unaffected by most dilute aqueous solutions, except ammonia and HF. It is not recommended for use with ethanol or organic solvents. The clear white markings make dark liquids easier to measure the the Teflon stopcock is less likely to freeze up.
Documents
Certificates
Search by Lot NumberSpecifications
- Description:Geissler Buret Locking Stopcock