Orders LinkContactUsLinkComponent
Rive Ray Box
Rive Ray Box
Five-Slit Screens for Light Source, Pack of 5
Catalog # 470211-170
Supplier #:  SV700
Rive Ray Box
Catalog # 470211-170
Rive Ray Box
Rive Ray Box
Five-Slit Screens for Light Source, Pack of 5
Price
$22.15
Log in for availability
Pack of 5 Case of 250
Product Details & Documents
High quality and low cost make this ideally suited for junior high or high school students.
  • Includes on/off switch
  • Slotted screen for multiple light rays
It is durably constructed with a rigid vinyl base and a metal box housing a transformer and a high intensity light. An adjustable screen produces either one wide beam or a choice of one, three, or five slit beams, all of which can be easily seen tracing a path across the table surface on which the unit sits.
Certifications: CSA approved.
Ordering information: Rive ray box is available as individual or case of 8; slit screen is available as a pack of 5 or case of 50 packs (250 ea.); and the replacement bulb is available as individual or case of 6.
Specifications
  • Description:
    Five-Slit Screens for Light Source, Pack of 5