VWR® Bin Liners
Gamma irradiated bin liner, Clear, LDPE, 40×48", 2 mil, Individually bagged and tagged in a double bagged pack
Supplier: VWR International
Jump to:
Product Details & Documents
LDPE liners for use in clean and controlled aseptic environments.
- Available non sterile or gamma irradiated
Uses include transport of product and waste disposal.
Specifications
- Color:Clear
- Description:Gamma irradiated bin liner
- Dimensions:40×48"
- Material:LDPE
- Thickness:2 mil
- Packaging:Individually bagged and tagged in a double bagged pack