Corning® Single-Use Bags for HYPERStack® Vessels, Corning
Quench bag, 5 L, ULDPE
Supplier: Corning
Supplier #: 91-200-76
Jump to:
Product Details & Documents
These single-use bags can be connected to tubing via tube welding or by using the pre-assembled multipurpose connectors.
They can be used to add media, trypsin, or quenching substrates to complete cell culture in an entirely closed solution.
Documents
Certificates
Search by Lot NumberPotential Applications and Scaling Methods of Extracellular Vesicles
Product Literature
Specifications
- Capacity:5 L
- Description:Quench bag
- Material:ULDPE
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.