Order Entry
Canada
Orders LinkContactUsLinkComponent
Phenol red solution indicator, Supelco®

Phenol red solution indicator, Supelco®

Glass bottle, 100 ml
Catalog # EM1.07242.0100
Supplier #:  1072420100
Phenol red solution indicator, Supelco®
Catalog # EM1.07242.0100
Supplier:  
CAS Number:  143-74-8
Phenol red solution indicator, Supelco®

Glass bottle, 100 ml

CAS Number:  143-74-8
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
PHENOL RED SOLN IN 100ML Catalog #CA1.07242.0100

Product CA1.07242.0100 is a replacement for EM1.07242.0100 and has the same packaging and specifications, but may be a different quantity.

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:143-74-8
  • Formula:C₁₉H₁₄O₅S
  • MDL Number:MFCD00003552
  • Molecular Weight:354.38 g/mol
  • Density:0.99 g/cm3 (20 °C)
  • Storage Temperature:Ambient

Documents

Specifications

Specification Test Results

  • Application test:
    conforms
  • pH-value (20 °C):
    7.0 - 7.3

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.