ɑ-Asarone
Product Details & Documents
Powder
CAS No.: 2883-98-9 Molecular Formula: C12H16O3 Formula Weight: 208.25 Physical Form: Powder Sensitivity: Light Sensitive Appearance: White to yellow Melting Point: 57-61° Boiling Point: 296° Beilstein Reference: 1910606 MDL No.: MFCD00064457
- CAS Number:2883-98-9
- Formula:C₁₂H₁₆O₃
- MDL Number:MFCD00064457
- Molecular Weight:208.26 g/mol
- Merck Index:14,824
- Boiling Pt:296 °C
- Melting Pt:57…61 °C
- Storage Temperature:Ambient
- EINECS:220-743-6
Documents
Certificates
Search by Lot NumberAlfa Aesar_Buy 3 Get 2 Complimentary items
Promotion Material
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Size:1 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



