LeStreak® Microstreakers, Decon Labs
Product Details & Documents
LeStreak® Microstreakers are used to inoculate or transfer microbiological samples
Small LeStreak® loop: Int.Ø 2.26 mm
- Medium LeStreak® loop: Int.Ø 2.91 mm
- Large LeStreak® loop: Int.Ø 3.57 mm
Decon’s LeStreak® microstreakers are loops made of high-quality, twisted nichrome wire that is permanently attached to lightweight, insulated handles which are perfectly balanced for easy manipulation. LeStreak® microstreakers slide smoothly across the agar surface with out tearing or gouging the medium. The nichrome wire cools rapidly after flaming.
The 1 and 10 µl (0.001 and 0.01 ml)) calibrated inoculating LeStreakers are individually calibrated in accordance with ASM/APHA via Evans blue dye test which is a CAP approved method. Calibration certificates are sent with calibrated loops or are available from Decon labs.
: UL listed.
Certificates
Search by Lot NumberSpecifications
- Capacity:1 µl (4 mm)
- Description:Calibrated streaker
- Handle color:White



