Order Entry
Canada
Orders LinkContactUsLinkComponent
H-Tyr(Tbu)-2-Chlorotrityl

H-Tyr(Tbu)-2-Chlorotrityl

5 g
Catalog # D-2890.0005BA
Supplier:   Bachem Americas
Supplier #:  4068311.0005
H-Tyr(Tbu)-2-Chlorotrityl
Catalog # D-2890.0005BA
Supplier:  Bachem Americas
CAS Number:  
H-Tyr(Tbu)-2-Chlorotrityl

5 g

CAS Number:  
Price
$1,247.83
Log in for availability
Each (5g)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Size:
    5 g
  • Cat. No.:
    D-2890.0005BA
  • Protein/peptide name:
    H-Tyr(Tbu)-2-Chlorotrityl
  • Storage conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD00801420
  • Sequence:
    H-Tyr(tBu)-2-chlorotrityl resin (200-400 mesh) (Low Substitution)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.